missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek ELx405 Select Deep Well Microplate Washer
GREENER_CHOICE

Product Code. p-4252752 Shop All Agilent BioTek Products
Click to view available options
Includes:
Deep well
Deep well, 4-Buffer Switching
Deep well, Ultrasonic Advantage
Deep well, Ultrasonic Advantage, 4-Buffer Switching
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17277979

Brand: Agilent BioTek ELX405SN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates.

The BioTek ELx405 Select is the first deep well plate washer that aspirates and dispenses into 96- and 384-well deep well and standard height microplates. The ELx405 Select is available in several configurations to address a wide variety of plate washing applications, including ELISA and cell-based assays.

Maintenance is easy with the available patented Ultrasonic Advantage that automates and cleans manifold tubes to remove built-up salt and protein.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Certifications

CE and ETL marked

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE and TUV marked. RoHS compliant.
Precision ≤3% CV
Plate Type 96-well Plate, 384-well Plate
Residual Volume ≤2 μL/well
Soak Time Programmable in minutes and seconds, up to 60 min.
Type Microplate Washer
Weight (Metric) 13.5 kg
Wash Cycles 1 to 10
Display Backlit LCD
Fluid Delivery Type One positive displacement pump
Humidity 10 to 80%, noncondensing
Frequency 50/60 Hz
Model ELX405UD
Temperature (English) Operating 59°F to 86°F
Temperature (Metric) Operating 15°C to 30°C
Includes Deep well
Width (English) 14 in.
Width (Metric) 35.6 cm
Height (English) 10 in.
Height (Metric) 25.4 cm
Depth (English) 17 in.
Depth (Metric) 43.2 cm
Weight (English) 30 lb.
Voltage 100/240 V
Amperage <8 A
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.