Learn More
Agilent BioTek ELx405 Qualification Package - IQ/OQ/PQ
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
ELx 405 Microplate Washer
Unit Size:
Each
Description
- Accessory for Agilent BioTek instruments range of microplate washers, readers and cell imagers
- Comes on USB flash drive with the following contents:
- Detailed required material list
- Installation qualification procedure and checklist
- Operational qualification procedure
- Performance qualification procedure
- Microsoft Excel spreadsheets for calculations required by procedures
- Maintenance procedures
- 5 year logbook for documenting all procedures/results
- All of the documentation is available in both .doc and .pdf file format
Note:
- Absorbance test plate (7260522) may be required in order to run some procedures specified for absorbance and multi-mode readers, but is not included and must be ordered separately.
Specifications
Specifications
| Includes | Detailed required material list, Installation qualification procedure and checklist, Operational qualification procedure, Performance qualification procedure, Microsoft Excel spreadsheets for calculations, Maintenance procedures, 5 year logbook |
| Type | Microplate Washer Accessories |
| For Use With (Equipment) | ELx 405 Microplate Washer |