missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELP4 Recombinant Rabbit Monoclonal Antibody (23GB5310), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | ELP4 |
|---|---|
| Clone | 23GB5310 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089539
![]() |
Invitrogen™
MA553882 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088179
|
Invitrogen™
MA553883 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELP4 Recombinant Monoclonal antibody specifically detects ELP4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| ELP4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q96EB1, Q9ER73 | |
| 26610, 687694, 77766 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5310 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A330107A17Rik; AN; C11orf19; dJ68P15A.1; elongation protein 4 homolog; elongation protein 4 homolog (S. cerevisiae); elongator acetyltransferase complex subunit 4; elongator complex protein 4; ELP4; hELP4; hypothetical protein LOC614139; PAX6 neighbor gene protein; PAX6NEB; PAXNEB; RGD1566052; similar to elongation protein 4 homolog | |
| A synthesized peptide derived from human ELP4 (313-364AA). | |
| Primary | |
| -20°C | |
| Elp4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title