missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ELP4 Recombinant Rabbit Monoclonal Antibody (23GB5310), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553883
This item is not returnable.
View return policy
Description
This gene encodes a component of the six subunit elongator complex, a histone acetyltransferase complex that associates directly with RNA polymerase II during transcriptional elongation. The human gene can partially complement sensitivity phenotypes of yeast ELP4 deletion mutants. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
Specifications
| ELP4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Elp4 | |
| A330107A17Rik; AN; C11orf19; dJ68P15A.1; elongation protein 4 homolog; elongation protein 4 homolog (S. cerevisiae); elongator acetyltransferase complex subunit 4; elongator complex protein 4; ELP4; hELP4; hypothetical protein LOC614139; PAX6 neighbor gene protein; PAX6NEB; PAXNEB; RGD1566052; similar to elongation protein 4 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26610, 687694, 77766 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5310 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96EB1, Q9ER73 | |
| Elp4 | |
| A synthesized peptide derived from human ELP4 (313-364AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction