missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1 Rabbit anti-Human, Mouse, Rat, Clone: 10U8X1, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Elk-1 |
|---|---|
| Clone | 10U8X1 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388311
|
Bio-Techne
NBP3-15631-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18394523
|
Novus Biologicals
NBP3-15631-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Elk-1 Monoclonal antibody specifically detects Elk-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| Elk-1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer, MAP Kinase Signaling, Neuroscience | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2002 | |
| IgG | |
| Affinity purified |
| 10U8X1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 329-428 of human Elk-1 (P19419). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title