missing translation for 'onlineSavingsMsg'
Learn More

Elk-1 Rabbit anti-Human, Mouse, Rat, Clone: 10U8X1, Novus Biologicals™

Product Code. 18394523 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394523 100 μg 100µL
18388311 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18394523 Supplier Novus Biologicals Supplier No. NBP315631100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Elk-1 Monoclonal antibody specifically detects Elk-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Elk-1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 10U8X1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 329-428 of human Elk-1 (P19419).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, MAP Kinase Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2002
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.