missing translation for 'onlineSavingsMsg'
Learn More

Elk-1, p Ser389 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£163.00 - £539.00

Specifications

Concentration 1 mg/mL
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Target Species Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18157809
View Documents
Novus Biologicals
NBP2-54759
0.1 mg
£539.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18211663
View Documents
Novus Biologicals
NBP2-54759-0.05MG
0.05 mg
£345.00
0.05mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18679716
View Documents
Novus Biologicals
NBP2-54759-0.025mg
0.025 mg
£163.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Elk-1 Polyclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

1 mg/mL
Unconjugated
Human
Elk1 (Phospho-Ser389) antibody was raised against a peptide sequence around phosphorylation site of serine 389 (P-R-S (p) -P-A) derived from Human Elk-1.
Primary
Polyclonal
Rabbit
ELK1
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.