missing translation for 'onlineSavingsMsg'
Learn More

Elk-1, p Ser389 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-54759-0.025mg

 View more versions of this product

Product Code. 18679716

  • £163.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Elk-1 Polyclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Elk1 (p Ser389)
Polyclonal
Unconjugated
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
ELK1
Elk1 (Phospho-Ser389) antibody was raised against a peptide sequence around phosphorylation site of serine 389 (P-R-S (p) -P-A) derived from Human Elk-1.
Epitope affinity purified
RUO
2002
Human
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
1 mg/mL
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:10-1:500
ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1
Rabbit
62 kDa
0.025 mg
Primary
This antibody detects endogenous level of Elk-1 only when phosphorylated at serine 389.
Store at -20°C. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.