missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1 Antibody (7E10D5), Novus Biologicals™
Mouse Monoclonal Antibody
£139.00
Specifications
| Antigen | Elk-1 |
|---|---|
| Clone | 7E10D5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
Description
Elk-1 Monoclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| Elk-1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Autophagy, Cancer, MAP Kinase Signaling | |
| Ascites with 0.03% Sodium Azide | |
| 2002 | |
| Ni-NTA purified truncated recombinant ELK1 expressed in E. Coli strain BL21 (DE3) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 7E10D5 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| ELK1 | |
| IgG1 | |
| Protein A purified | |
| ELK1 (7E10D5) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title