missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Elk-1 Antibody (7E10D5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60513-0.025mg
This item is not returnable.
View return policy
Description
Elk-1 Monoclonal specifically detects Elk-1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Elk-1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| ELK1 (7E10D5) | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 7E10D5 | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| ELK1 | |
| Ni-NTA purified truncated recombinant ELK1 expressed in E. Coli strain BL21 (DE3) | |
| 0.025 mg | |
| Autophagy, Cancer, MAP Kinase Signaling | |
| 2002 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction