missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF3/ESE-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£163.00 - £367.00
Specifications
| Antigen | ELF3/ESE-1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18259581
|
Novus Biologicals
NB600-212SS |
0.025 mL |
£163.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18254012
|
Novus Biologicals
NB600-212 |
0.1 mL |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELF3/ESE-1 Polyclonal specifically detects ELF3/ESE-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ELF3/ESE-1 | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Transcription Factors and Regulators | |
| PBS with 0.02% Sodium Azide | |
| 1999 | |
| N-terminal sequence and C-terminal sequence of the human ESE-1 protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Human | |
| E74-like factor 3, E74-like factor 3 (ets domain transcription factor, epithelial-specific ), Epithelial-restricted with serine box, epithelial-specific), Epithelium-restricted Ets protein ESX, Epithelium-specific Ets transcription factor 1, EPR-1, ets domain transcription factor, serine box (epithelial-specific), ETS-related transcription factor Elf-3, JEN, serine box | |
| ELF3 | |
| IgG | |
| Affinity Purified | |
| This recognizes both isoforms of ESE-1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title