missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELF3/ESE-1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB600-212
This item is not returnable.
View return policy
Description
ELF3/ESE-1 Polyclonal specifically detects ELF3/ESE-1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ELF3/ESE-1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ELF3 | |
| N-terminal sequence and C-terminal sequence of the human ESE-1 protein | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Transcription Factors and Regulators | |
| 1999 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:500, Immunocytochemistry/Immunofluorescence | |
| E74-like factor 3, E74-like factor 3 (ets domain transcription factor, epithelial-specific ), Epithelial-restricted with serine box, epithelial-specific), Epithelium-restricted Ets protein ESX, Epithelium-specific Ets transcription factor 1, EPR-1, ets domain transcription factor, serine box (epithelial-specific), ETS-related transcription factor Elf-3, JEN, serine box | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This recognizes both isoforms of ESE-1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction