missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Reactive Oxygen Species (ROS) Fluorometric Assay Kit

Product Code. 17932171
Change view
Click to view available options
Quantity:
96 tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932171 96 tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17932171 Supplier Invitrogen™ Supplier No. EEA019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.

The Reactive Oxygen Species (ROS) Fluorometric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of ROS in fresh tissue and live cell samples. ROS are highly reactive molecules that play a crucial role in various physiological and pathological processes, including oxidative stress, inflammation, and cellular signaling.

The assay works by utilizing the fluorescent probe 2',7'-dichlorodihydrofluorescein diacetate (DCFH-DA) to detect the presence of ROS. DCFH-DA is taken up by cells and is converted to dichlorofluorescein (DCFH) by intracellular esterase. DCFH is oxidized by ROS to the strong fluorescent DCF. The fluorescence of DCF (Ex/Em 502/525 nm) is directly proportional to the amount of ROS present in the sample.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Reagent 1: –20°C
Reagent 2: 2–8°C

Detection Method Fluorescence
Sample Type Cell Culture, Live Tissue
For Use With (Equipment) Spectrophotometer, Microplate Reader, Flow Cytometer
Incubation Time 30-60 min.
Quantity 96 tests
Shipping Condition Dry Ice, 2–8°C
Target Kit Named Reactive Oxygen Species (ROS)
Total Assay Time 60 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.