missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek EL406 Microplate Washer/Dispenser
GREENER_CHOICE

Product Code. 17277889 Shop All Agilent BioTek Products
Click to view available options
Model:
406PF
406PSNA4
406PSU0LB
406PSU0SB
406PSUB1
406PSUB1LB
406PSUB1SB
406PSUB2LB
406PSUB2SB
406PSUB3
406PSUB3LB
406PUB1
406PUB3
406SNA0LB
406SNA4
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17277889

Brand: Agilent BioTek 406PSU0LBSN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The BioTek EL406 washer dispenser is a compact instrument offering fast, full plate washing along with three reagent dispensers.

The BioTek EL406 washer dispenser is a compact instrument offering fast, full plate washing along with three reagent dispensers. The EL406 capabilities include washing and dispensing for standard assays, plus biomagnetic separation and vacuum filtration protocols.

The Dual-Action manifold optimizes washing for loosely adherent cell monolayers. The patented Ultrasonic Advantage prevents blockages in the manifold tubes before washing, to reduce assay failure. This modular plate washer dispenser offers flexibility with both syringe pump and peristaltic pump dispensers, along with plate washing, on a single platform.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Certifications

CE marked; ETL listed

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±3%; Dispensing: ±1 μL at 5 μL, +1 μL at 20 μL, ±1% at 100 μL
Certifications/Compliance CE and TUV marked. RoHS compliant.
Bottles Optional (1 or 2 L)
Precision Wash: <3% CV (model dependent); Dispense (Syringe Pump): ≤5% CV at 5 μL, ≤2.5% CV at 20 μL, ≤1% CV at 100 μL
Dispense Volume 500 nL to 3000 μL/well
Plate Type 96-, 384- and 1536-well; Low profile and standard height; Solid and filter bottom (option) plates
Residual Volume ≤2 μL/well
Washing Speed 96 wells: 300 μL/well, 96-tube manifold: 13 s; 384 wells: 100 μL/well, 192-tube manifold: 17 s; 1536 wells: 10 μL/wells, two 32-tube manifolds: 36 s
Soak Time Programmable in minutes and seconds, up to 60 min.
Type Microplate Washer/Dispenser
Weight (Metric) 14.5 kg
Wash Cycles 1 to 250
Flow Rate High to Low Flow; Optimized rates for cell assays
Frequency 50/60 Hz
Model 406PSU0LB
Increments 1 μL
Includes 1536-well microplate washing and dispensing (large bore, steel tubes); 1-Reagent Dispense Module – 1 Peristaltic Pump (autoclavable, 8-tip (1 x 8) cassette/sapphire jeweled, steel or plastic tips); 2-Reagent Dispense Module – 2 Syringe Pumps (autoclavable/(2) 16-tube (1 x 16) manifolds/steel tubes); Ultrasonic Advantage
Width (English) 16.5 in.
Width (Metric) 42 cm
Height (English) 12.5 in.
Height (Metric) 31.8 cm
Depth (English) 18 in.
Depth (Metric) 46 cm
Weight (English) 32 lb.
Voltage 100/240 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.