Learn More
Agilent BioTek EL406 Dual Syringe Dispenser Automatic Reagent Switching Module
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
EL406 Microplate Washer, Agilent BioTek Microplate Washer and Reader Instruments
Unit Size:
Each
Description
- Adds additional two reagents to an EL406's syringe dispenser capabilities or provides a rinse solution for maintaining each of the dual syringe module's reagent fluid lines
Specifications
Specifications
| Type | Reagent Switching Module |
| For Use With (Equipment) | EL406 Microplate Washer, Agilent BioTek Microplate Washer and Reader Instruments |