missing translation for 'onlineSavingsMsg'
Learn More

EIF4H Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen EIF4H
Concentration 0.3 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16807793
View Documents
Proteintech
11012-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16897783
View Documents
Proteintech
11012-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EIF4H has two isoforms. The short isoform (∽25kDa) is the predomit isoform and is expressed alone in liver and skeletal muscle. This is a rabbit anti-EIF4H polyclonal antibody raised against part of N-terminal chain of human EIF4H, and can recognize

This gene encodes one of the translation initiation factors, which functions to stimulate the initiation of protein synthesis at the level of mRNA utilization. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.
Specifications

Specifications

EIF4H
Western Blot
Unconjugated
Rabbit
Human
Q15056
7458
EIF4H Fusion Protein Ag1468
Primary
-20°C
EIF4H
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
eIF 4H, EIF4H, KIAA0038, WBSCR1, WSCR1
EIF4H
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.