missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4H Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11012-1-AP-20UL
This item is not returnable.
View return policy
Description
EIF4H has two isoforms. The short isoform (∽25kDa) is the predomit isoform and is expressed alone in liver and skeletal muscle. This is a rabbit anti-EIF4H polyclonal antibody raised against part of N-terminal chain of human EIF4H, and can recognize
This gene encodes one of the translation initiation factors, which functions to stimulate the initiation of protein synthesis at the level of mRNA utilization. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.Specifications
| EIF4H | |
| Polyclonal | |
| Unconjugated | |
| EIF4H | |
| eIF 4H, EIF4H, KIAA0038, WBSCR1, WSCR1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7458 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15056 | |
| EIF4H | |
| EIF4H Fusion Protein Ag1468 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction