missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£167.00 - £302.00
Specifications
| Antigen | eIF4A1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.25-1.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18126921
|
Novus Biologicals
NBP2-24632SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18197681
|
Novus Biologicals
NBP2-24632 |
0.1 mg |
£302.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF4A1 Polyclonal specifically detects eIF4A1 in Human, Mouse, Rat, Canine, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| eIF4A1 | |
| Western Blot 0.25-1.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Canine, Chicken, Primate | |
| P60842 | |
| 1973 | |
| A portion of amino acid 350-400 of human eIF4A1 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| ATP-dependent RNA helicase eIF4A-1, DDX2Aeukaryotic initiation factor 4A-I, EC 3.6.1, EC 3.6.4.13, EIF4A, EIF-4A, eIF-4A-I, eIF4A-I, eukaryotic initiation factor 4AI, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A, isoform 1, eukaryotic translation initiation factor 4A1 | |
| EIF4A1 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, dog, chicken, chimpanzee and monkey and 88% homologous in zebrafish. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title