missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-24632
This item is not returnable.
View return policy
Description
eIF4A1 Polyclonal specifically detects eIF4A1 in Human, Mouse, Rat, Canine, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| eIF4A1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| ATP-dependent RNA helicase eIF4A-1, DDX2Aeukaryotic initiation factor 4A-I, EC 3.6.1, EC 3.6.4.13, EIF4A, EIF-4A, eIF-4A-I, eIF4A-I, eukaryotic initiation factor 4AI, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A, isoform 1, eukaryotic translation initiation factor 4A1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, dog, chicken, chimpanzee and monkey and 88% homologous in zebrafish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 0.25-1.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| P60842 | |
| EIF4A1 | |
| A portion of amino acid 350-400 of human eIF4A1 was used as the immunogen. | |
| 0.1 mg | |
| Core ESC Like Genes, mTOR Pathway, Stem Cell Markers | |
| 1973 | |
| Human, Mouse, Rat, Canine, Chicken, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction