Learn More
Invitrogen™ Human EIF2AK3 (PERK), GST Tag Recombinant Protein
Human EIF2AK3 (PERK), GST Tag Recombinant Protein
Brand: Invitrogen™ PV5107
Code : NEW
Additional Details : Weight : 0.01000kg
Description
EIF2AK3 (PERK) phosphorylates Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER.
Specifications
NP_004827.3 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
EIF2AK3 (PERK) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
93.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
PEK Kinase Family | |
Recombinant | |
Baculovirus |