missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EIF2AK3 (PERK), GST Tag Recombinant Protein
Human EIF2AK3 (PERK), GST Tag Recombinant Protein
£440.00 - £6900.00
Specifications
Accession Number | NP_004827.3 |
---|---|
Gene Alias | EIF2AK3 (PERK) |
KinaseFamily | PEK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438703
|
Invitrogen™
PR8985A |
100μg |
£1604.00
100µg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488773
|
Invitrogen™
PV5107 |
1mg |
£6900.00
1mg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10698863
|
Invitrogen™
PV5106 |
10 μg |
£440.00
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
EIF2AK3 (PERK) phosphorylates Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER.
Specifications
NP_004827.3 | |
PEK Kinase Family | |
Recombinant | |
Baculovirus |
EIF2AK3 (PERK) | |
Human | |
GST Tag |