missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EIF2AK3 (PERK), GST Tag Recombinant Protein
Human EIF2AK3 (PERK), GST Tag Recombinant Protein
Brand: Invitrogen™ PV5106
Code : 50
Additional Details : Weight : 0.50000kg
Description
EIF2AK3 (PERK) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER.Specifications
NP_004827.3 | |
9451 | |
10 μg | |
PEK Kinase Family | |
Human | |
GST Tag | |
Biochemical terbium phospho-specific activity | |
Dry Ice | |
EIF2AK3 | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
93.6 kDa | |
EIF2AK3 (PERK) | |
aa 536-1116 | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: Other | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate | |
eukaryotic translation initiation factor 2-alpha kinase 3 | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |