missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2A, p Ser51 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£170.00 - £415.00
Specifications
| Antigen | eIF2A (p Ser51) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18211195
|
Novus Biologicals
NB100-81896-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18767253
|
Novus Biologicals
NB100-81896 |
£357.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18527790
|
Novus Biologicals
NB100-81896-0.1mg |
0.1 mg |
£415.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF2A Polyclonal specifically detects eIF2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| eIF2A (p Ser51) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| DNA Repair, Phospho Specific, Translation Control, Unfolded Protein Response | |
| P05198 | |
| 83939 | |
| IgG | |
| Affinity Purified |
| 1.0 mg/mL | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse, Rat, Human | |
| EIF-2, EIF-2A, EIF2AEIF2, EIF-2alpha, eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa, subunit 1 (alpha, 35kD ) | |
| EIF2A | |
| Primary | |
| 38 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title