missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2A, p Ser51 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-81896-0.1mg
This item is not returnable.
View return policy
Description
eIF2A Polyclonal specifically detects eIF2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| eIF2A (p Ser51) | |
| Polyclonal | |
| Unconjugated | |
| phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
| EIF-2, EIF-2A, EIF2AEIF2, EIF-2alpha, eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa, subunit 1 (alpha, 35kD ) | |
| Rabbit | |
| 38 kDa | |
| 0.1 mg | |
| DNA Repair, Phospho Specific, Translation Control, Unfolded Protein Response | |
| 83939 | |
| Mouse, Rat, Human | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
| P05198 | |
| EIF2A | |
| The antiserum was produced against synthesized phosphopeptide derived from human eIF2A around the phosphorylation site of serine 51 (E-L-SP-R-R). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects endogenous levels of eIF2A only when phosphorylated at serine 51. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction