missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EHMT2 Recombinant Rabbit Monoclonal Antibody (23GB825), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | EHMT2 |
|---|---|
| Clone | 23GB825 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088203
![]() |
Invitrogen™
MA554098 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088583
|
Invitrogen™
MA554099 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EHMT2 Recombinant Monoclonal antibody specifically detects EHMT2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| EHMT2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q96KQ7, Q9Z148 | |
| 10919, 110147, 361798 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB825 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ankyrin repeat-containing protein; Bat8; C6orf30; D17Ertd710e; DKFZp686H08213; Ehmt2; euchromatic histone lysine methyltransferase 2; euchromatic histone lysine N-methyltransferase 2; euchromatic histone-lysine N-methyltransferase 2; FLJ35547; G9a; G9A histone methyltransferase; GAT8; H3 ly; H3-K9-Histone-Methyl-Transferase; H3-K9-HMTase 3; histone H3-K9 methyltransferase 3; Histone-lysine N-methyltransferase EHMT2; histone-lysine N-methyltransferase, H3 lysine-9 specific 3; HLA-B associated transcript 8; HLA-B associated transcript 8, rat orthologue; HLA-B-associated transcript 8; KMT1C; lysine N-methyltransferase 1C; NG36; NG36/G9a; Protein G9a | |
| A synthetic peptide of human EHMT2 (2-300AA). | |
| Primary | |
| -20°C | |
| Ehmt2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title