missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EHMT2 Recombinant Rabbit Monoclonal Antibody (23GB825), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554098
This item is not returnable.
View return policy
Description
A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. This gene is found near this cluster; it was mapped near the gene for C2 within a 120-kb region that included a HSP70 gene pair. These genes are all within the human major histocompatibility complex class III region. This gene was thought to be two different genes, NG36 and G9a, adjacent to each other but a recent publication shows that there is only a single gene. The protein encoded by this gene is thought to be involved in intracellular protein-protein interaction. There are three alternatively spliced transcript variants of this gene but only two are fully described.
Specifications
| EHMT2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Ehmt2 | |
| ankyrin repeat-containing protein; Bat8; C6orf30; D17Ertd710e; DKFZp686H08213; Ehmt2; euchromatic histone lysine methyltransferase 2; euchromatic histone lysine N-methyltransferase 2; euchromatic histone-lysine N-methyltransferase 2; FLJ35547; G9a; G9A histone methyltransferase; GAT8; H3 ly; H3-K9-Histone-Methyl-Transferase; H3-K9-HMTase 3; histone H3-K9 methyltransferase 3; Histone-lysine N-methyltransferase EHMT2; histone-lysine N-methyltransferase, H3 lysine-9 specific 3; HLA-B associated transcript 8; HLA-B associated transcript 8, rat orthologue; HLA-B-associated transcript 8; KMT1C; lysine N-methyltransferase 1C; NG36; NG36/G9a; Protein G9a | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10919, 110147, 361798 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB825 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96KQ7, Q9Z148 | |
| Ehmt2 | |
| A synthetic peptide of human EHMT2 (2-300AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction