missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EHD1 Recombinant Rabbit Monoclonal Antibody (23GB5280), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | EHD1 |
|---|---|
| Clone | 23GB5280 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087764
![]() |
Invitrogen™
MA552830 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087469
|
Invitrogen™
MA552831 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EHD1 Recombinant Monoclonal antibody specifically detects EHD1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| EHD1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q641Z6, Q9H4M9, Q9WVK4 | |
| 10938, 13660, 293692 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5280 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AA409636; CDABP0131; EH domain containing 1; EH domain-containing protein 1; Ehd1; EH-domain containing 1; H-PAST; HPAST1; mPAST1; PAST; PAST homolog 1; PAST1; RGD1306960; RME-1; testilin | |
| A synthesized peptide derived from human EHD1 (1-45AA). | |
| Primary | |
| -20°C | |
| EHD1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title