Learn More
Invitrogen™ EHD1 Recombinant Rabbit Monoclonal Antibody (23GB5280), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA552830
 View more versions of this product
Product Code. 30087764
  • £387.00 / 100µL
Description
EHD1 acts in early endocytic membrane fusion and membrane trafficking of recycling endosomes. EHD1 also plays a role in myoblast fusion and is recruited to endosomal membranes upon nerve growth factor stimulation, indirectly regulates neurite outgrowth. Further, EHD1 belongs to a highly conserved gene family encoding EPS15 homology (EH) domain-containing proteins. The protein-binding EH domain was first noted in EPS15, a substrate for the epidermal growth factor receptor. The EH domain has been shown to be an important motif in proteins involved in protein-protein interactions and in intracellular sorting. The EHD1 protein is thought to play a role in the endocytosis of IGF1 receptors. Alternatively spliced transcript variants have been found for the EHD1 gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EHD1
Recombinant Monoclonal
Unconjugated
EHD1
AA409636; CDABP0131; EH domain containing 1; EH domain-containing protein 1; Ehd1; EH-domain containing 1; H-PAST; HPAST1; mPAST1; PAST; PAST homolog 1; PAST1; RGD1306960; RME-1; testilin
Rabbit
Affinity chromatography
RUO
10938, 13660, 293692
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB5280
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q641Z6, Q9H4M9, Q9WVK4
EHD1
A synthesized peptide derived from human EHD1 (1-45AA).
100 μL
Primary
Human, Mouse, Rat