missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN1/PHD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 74 publications
£491.00
Specifications
| Antigen | EGLN1/PHD2 |
|---|---|
| Concentration | 1 mg/mL |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
EGLN1/PHD2 Polyclonal specifically detects EGLN1/PHD2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| EGLN1/PHD2 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Hypoxia | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179 | |
| EGLN1 | |
| IgG | |
| Affinity Purified | |
| Human. This antibody does not appear to work in Mouse. Results have been mixed in Rat with success in western analysis and immunofluorescence on Rat endothelial cells and negative results with PC12 cells. Expected reactivity with Rabbit, Orangutan, Rhesus Monkey, Gorilla based on 100% sequence identity. |
| 1 mg/mL | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| Q9GZT9 | |
| 54583 | |
| The epitope recognized by this antibody maps to a region between residues 1 and 50 of human PHD2/HIF Prolyl Hydroxylase 2 using the numbering given in entry NP_071334.1 (GeneID 54583). | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 46 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title