missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN1/PHD2 Antibody (366G/76/3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
£173.00 - £382.00
Specifications
| Antigen | EGLN1/PHD2 |
|---|---|
| Clone | 366G/76/3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-2.0 ug/ml, Simple Western 1:50, Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18079113
|
Novus Biologicals
NBP1-30328SS |
0.025 mL |
£173.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18775123
|
Novus Biologicals
NBP1-30328 |
0.1 mL |
£382.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGLN1/PHD2 Monoclonal specifically detects EGLN1/PHD2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| EGLN1/PHD2 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Hypoxia | |
| Q9GZT9 | |
| 54583 | |
| A peptide within residues 1-50 of human PHD2/HIF Prolyl Hydroxylase 2. [Swiss-Prot# Q9GZT9] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 46 kDa |
| 366G/76/3 | |
| Western Blot 0.5-2.0 ug/ml, Simple Western 1:50, Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179 | |
| EGLN1 | |
| IgG1 | |
| Protein G purified | |
| Human. Immunogen sequence has 83% identity with mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title