missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGLN1/PHD2 Antibody (366G/76/3), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP1-30328SS
This item is not returnable.
View return policy
Description
EGLN1/PHD2 Monoclonal specifically detects EGLN1/PHD2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| EGLN1/PHD2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5-2.0 ug/ml, Simple Western 1:50, Immunohistochemistry 1:400, Immunohistochemistry-Paraffin 1:400 | |
| C1orf12HPH2, EC 1.14.11, EC 1.14.11.-, ECYT3, egl nine homolog 1, egl nine homolog 1 (C. elegans), egl nine-like protein 1, HIF prolyl hydroxylase 2, HIF-PH2, HIFPH2 EGL nine (C.elegans) homolog 1, HIF-prolyl hydroxylase 2, HPH-2, Hypoxia-inducible factor prolyl hydroxylase 2, PHD2SM20, Prolyl hydroxylase domain-containing protein 2, SM-20, zinc finger MYND domain-containing protein 6, ZMYND6DKFZp761F179 | |
| Mouse | |
| 46 kDa | |
| 0.025 mL | |
| Cancer, Hypoxia | |
| 54583 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 366G/76/3 | |
| Unconjugated | |
| Q9GZT9 | |
| EGLN1 | |
| A peptide within residues 1-50 of human PHD2/HIF Prolyl Hydroxylase 2. [Swiss-Prot# Q9GZT9] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human. Immunogen sequence has 83% identity with mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction