missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EGFR (ErbB1) [T790M, L858R], GST Tag Recombinant Protein
Human EGFR (ErbB1) [T790M, L858R], GST Tag Recombinant Protein
£435.00
Specifications
Accession Number | NP_005219.2; T790M L858R |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 90.6 kDa |
Gene Alias | EGFR (ErbB1) T790M L858R |
KinaseFamily | EGF Receptor (ErbB) Kinase Family |
Description
The epidermal growth factor receptor is a receptor tyrosine kinases that signals in response to various growth factors. Overexpression has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. The double T790M L858R mutant of EGFR displays resistance to Erlotinib (Tarceva) and sensitivity to HKI-272 and rapamycin combination therapy.Specifications
NP_005219.2; T790M L858R | |
90.6 kDa | |
EGF Receptor (ErbB) Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Polysorbate 20, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice |
Kinase Assay | |
EGFR (ErbB1) T790M L858R | |
Human | |
GST Tag | |
Tyrosine Kinases | |
Baculovirus |