missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGF Antibody (18B12), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | EGF |
|---|---|
| Clone | 18B12 |
| Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043216
|
Novus Biologicals
NBP3-26606-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043728
|
Novus Biologicals
NBP3-26606-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EGF Monoclonal antibody specifically detects EGF in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
| EGF | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG | |
| A synthesized peptide derived from Human EGF [UniProt P01133] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 18B12 | |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1950 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title