missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EGF Antibody (18B12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26606-50ul
This item is not returnable.
View return policy
Description
EGF Monoclonal antibody specifically detects EGF in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| EGF | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry | |
| 18B12 | |
| Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| beta-urogastrone, epidermal growth factor, epidermal growth factor (beta-urogastrone), hEGF, HOMG4, pro-epidermal growth factor, URG | |
| A synthesized peptide derived from Human EGF [UniProt P01133] | |
| 50 μL | |
| Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication, Cytokine Research, Immunology, Virology Bacteria and Parasites | |
| 1950 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering