missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFCAB4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-55340
This item is not returnable.
View return policy
Description
EFCAB4B Polyclonal specifically detects EFCAB4B in Human samples. It is validated for Western Blot.
Specifications
| EFCAB4B | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| Calcium release-activated calcium channel regulator 2A, CRAC channel regulator 2A, CRACR2A, DKFZp686G13246, EF-hand calcium binding domain 4B, EF-hand calcium-binding domain-containing protein 4B, FLJ33046, FLJ33805, MGC4266 | |
| Rabbit | |
| 45 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9BSW2 | |
| CRACR2A | |
| Synthetic peptides corresponding to EFCAB4B(EF-hand calcium binding domain 4B) The peptide sequence was selected from the middle region of EFCAB4B. Peptide sequence KNELECALKRKIAAYDEEIQHLYEEMEQQIKSEKEQFLLKDTERFQARSQ. | |
| Affinity purified | |
| RUO | |
| 84766 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction