missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFCAB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57043
This item is not returnable.
View return policy
Description
EFCAB3 Polyclonal specifically detects EFCAB3 in Human samples. It is validated for Western Blot.
Specifications
| EFCAB3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EF-hand calcium binding domain 3, EF-hand calcium-binding domain-containing protein 3, FLJ25818, MGC126801, MGC126827 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 146779 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8N7B9 | |
| EFCAB3 | |
| Synthetic peptides corresponding to EFCAB3 (EF-hand calcium binding domain 3) The peptide sequence was selected from the N terminal of EFCAB3. Peptide sequence MAVSEIKPKLKLNPLTKVPISHNKRDRDLPGSLQCQLQHKEKKLSASQMA. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%. | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction