missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EEF2K Recombinant Human Protein
EEF2K is a highly conserved serine/threoning kinase in the calmodulin signaling pathway.
Brand: Invitrogen™ PV4560
Code : NEW
Additional Details : Weight : 0.01000kg
Description
EEF2K activity is increased in cancers, including breast cancer.
Specifications
NP_037434 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
109.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
Alpha Kinase Family | |
Recombinant | |
E. coli |
Nuclear Receptor Assay | |
29904 | |
1mg | |
RUO - research use only | |
EEF2K, eEF-2K | |
Human | |
GST Tag | |
Frozen |