missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EEF2K Recombinant Human Protein
EEF2K is a highly conserved serine/threoning kinase in the calmodulin signaling pathway.
£434.00 - £6900.00
Specifications
Accession Number | NP_037434 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Gene Alias | EEF2K, eEF-2K |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10046532
|
Invitrogen™
PV4559 |
10μg |
£434.00
10µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15458673
|
Invitrogen™
PR8535A |
100μg |
£1602.00
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15418733
|
Invitrogen™
PV4560 |
1mg |
£6900.00
1mg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
EEF2K activity is increased in cancers, including breast cancer.
Specifications
NP_037434 | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
EEF2K, eEF-2K | |
Human | |
GST Tag |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
RUO - research use only | |
Alpha Kinase Family | |
Recombinant | |
E. coli |