missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF2K Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | EEF2K |
|---|---|
| Concentration | 0.35 mg/mL |
| Applications | Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16821764
|
Proteintech
13510-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16811764
|
Proteintech
13510-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD54, M.W. 90,000, antigen is the Intercellular Adhesion Molecule-1 (ICAM-1) and is the ligand for LFA-1 mediated adhesion. CD54 is weakly expressed on peripheral blood leukocytes. Upon activation the antigen is expressed strongly on B cells, T cells, macrophages and granulocytes. ICAM-1 is also inducible on endothelial cells, epithelial cells and fibroblasts.Specifications
| EEF2K | |
| Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| O00418, O08796, P70531 | |
| 13631, 25435, 29904 | |
| EEF2K Fusion Protein Ag4432 | |
| Primary | |
| -20°C | |
| EEF2K |
| 0.35 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| eEF 2 kinase, eEF 2K, EEF2K, HSU93850 | |
| Eef2k | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title