missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF2K Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13510-1-AP-150UL
This item is not returnable.
View return policy
Description
CD54, M.W. 90,000, antigen is the Intercellular Adhesion Molecule-1 (ICAM-1) and is the ligand for LFA-1 mediated adhesion. CD54 is weakly expressed on peripheral blood leukocytes. Upon activation the antigen is expressed strongly on B cells, T cells, macrophages and granulocytes. ICAM-1 is also inducible on endothelial cells, epithelial cells and fibroblasts.Specifications
| EEF2K | |
| Polyclonal | |
| Unconjugated | |
| EEF2K | |
| eEF 2 kinase, eEF 2K, EEF2K, HSU93850 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13631, 25435, 29904 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00418, O08796, P70531 | |
| Eef2k | |
| EEF2K Fusion Protein Ag4432 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction