missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1B2 Mouse anti-Human, Clone: 3E1C11, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | EEF1B2 |
|---|---|
| Clone | 3E1C11 |
| Concentration | 1 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16887425
|
Proteintech
60329-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16877425
|
Proteintech
60329-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a translation elongation factor. The protein is a guanine nucleotide exchange factor involved in the transfer of aminoacylated tRNAs to the ribosome. Alternative splicing results in three transcript variants which differ only in the 5′ UTR.Specifications
| EEF1B2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| EEF1B, EEF1B1, EEF1B2, EF 1 beta, EF1B, Elongation factor 1 beta | |
| EEF1B2 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 3E1C11 | |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| P24534 | |
| 1933 | |
| EEF1B2 Fusion Protein Ag0135 | |
| Primary | |
| -20°C | |
| EEF1B2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title