missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EEF1B2 Mouse anti-Human, Clone: 3E1C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60329-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a translation elongation factor. The protein is a guanine nucleotide exchange factor involved in the transfer of aminoacylated tRNAs to the ribosome. Alternative splicing results in three transcript variants which differ only in the 5′ UTR.Specifications
| EEF1B2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24534 | |
| EEF1B2 | |
| EEF1B2 Fusion Protein Ag0135 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 3E1C11 | |
| Unconjugated | |
| EEF1B2 | |
| EEF1B, EEF1B1, EEF1B2, EF 1 beta, EF1B, Elongation factor 1 beta | |
| Mouse | |
| Protein G | |
| RUO | |
| 1933 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction