missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eEF1A1 Mouse anti-Human, Clone: 23C11, Novus Biologicals™
Mouse Monoclonal Antibody
£241.00 - £378.00
Specifications
| Antigen | eEF1A1 |
|---|---|
| Clone | 23C11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:3000, Flow Cytometry 1:10-1:1021, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18257121
|
Novus Biologicals
NBP2-59411-50UL |
50 μL |
£241.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18266931
|
Novus Biologicals
NBP2-59411-100UL |
100 μL |
£378.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eEF1A1 Monoclonal specifically detects eEF1A1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| eEF1A1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| CCS3, CCS-3, cervical cancer suppressor 3, CTCL tumor antigen, EE1A1, EEF-1, EEF1A, eEF1A-1, EF1AHNGC:16303, EF1a-like protein, EF-1-alpha-1, EF-Tu, elongation factor 1 alpha subunit, elongation factor 1-alpha 1, Elongation factor Tu, Eukaryotic elongation factor 1 A-1, eukaryotic translation elongation factor 1 alpha 1, eukaryotic translation elongation factor 1 alpha 1-like 14, FLJ25721, glucocorticoid receptor AF-1 specific elongation factor, GRAF-1EF, LENG7, leukocyte receptor cluster (LRC) member 7, Leukocyte receptor cluster member 7, MGC102687, MGC131894, MGC16224, prostate tumor-inducing protein 1, PTI1, translation elongation factor 1 alpha 1-like 14 | |
| EEF1A1 | |
| IgG2a κ | |
| Protein A purified |
| 23C11 | |
| Western Blot 1:3000, Flow Cytometry 1:10-1:1021, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 1915 | |
| Recombinant human EEF1A1 (1-462aa) purified from E.coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title