missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eEF1A1 Mouse anti-Human, Clone: 23C11, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59411-100UL
This item is not returnable.
View return policy
Description
eEF1A1 Monoclonal specifically detects eEF1A1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| eEF1A1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:3000, Flow Cytometry 1:10-1:1021, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| CCS3, CCS-3, cervical cancer suppressor 3, CTCL tumor antigen, EE1A1, EEF-1, EEF1A, eEF1A-1, EF1AHNGC:16303, EF1a-like protein, EF-1-alpha-1, EF-Tu, elongation factor 1 alpha subunit, elongation factor 1-alpha 1, Elongation factor Tu, Eukaryotic elongation factor 1 A-1, eukaryotic translation elongation factor 1 alpha 1, eukaryotic translation elongation factor 1 alpha 1-like 14, FLJ25721, glucocorticoid receptor AF-1 specific elongation factor, GRAF-1EF, LENG7, leukocyte receptor cluster (LRC) member 7, Leukocyte receptor cluster member 7, MGC102687, MGC131894, MGC16224, prostate tumor-inducing protein 1, PTI1, translation elongation factor 1 alpha 1-like 14 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1915 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 23C11 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| EEF1A1 | |
| Recombinant human EEF1A1 (1-462aa) purified from E.coli | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction