Learn More
3M™ Edge™ 5 Personal Noise Dosimeter
Product Code. 15719667 Shop All 3M Products
Change view
Click to view available options
Battery Life:
40 hours
Unit Size:
Each
Product Code. Battery Life Unit Size
15719667 40 hours Each
1 options
Product Code. 15719667 Supplier 3M™ Supplier No. 7000030071
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Edge™ 5 Personal Noise Dosimeter offers advanced technology packaged in a cable-free, compact frame for a lighter, more ergonomic way to monitor noise levels.
  • Rechargeable lithium polymer battery
  • Thin, contoured mount hugs the shoulder; compact frame offers superior comfort
  • Intrinsic safety certifications
  • Shoulder Mount
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 40 hours
Certifications/Compliance CE, IEC 61252, ANSI S1.25
Dimensions 88 x 53 x 19 mm
Includes Docking Station
Measurement Range Dose, Exposure, Identity Field, L(C-A), Lavg/Leq, Max, Min, Peak, Projected Dose, Run Time, SEL (Lep), SPL, Threshold, TWA, Upper Limit (UL)