missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDAR Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | EDAR |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Western Blot, Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16878064
|
Proteintech
18032-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16868064
|
Proteintech
18032-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal domit and recessive forms of hypohidrotic ectodermal dysplasia.Specifications
| EDAR | |
| Western Blot, Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9UNE0 | |
| 10913 | |
| EDAR Fusion Protein Ag12601 | |
| Primary | |
| -20°C | |
| EDAR |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DL, Downless homolog, Ectodermal dysplasia receptor, ectodysplasin A receptor, ED1R, ED3, ED5, EDA A1 receptor, EDA A1R, EDA1R, EDA3, EDAR, HRM1 | |
| EDAR | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title