missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDAR Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18032-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the tumor necrosis factor receptor family. The encoded transmembrane protein is a receptor for the soluble ligand ectodysplasin A, and can activate the nuclear factor-kappaB, JNK, and caspase-independent cell death pathways. It is required for the development of hair, teeth, and other ectodermal derivatives. Mutations in this gene result in autosomal domit and recessive forms of hypohidrotic ectodermal dysplasia.Specifications
| EDAR | |
| Polyclonal | |
| Unconjugated | |
| EDAR | |
| DL, Downless homolog, Ectodermal dysplasia receptor, ectodysplasin A receptor, ED1R, ED3, ED5, EDA A1 receptor, EDA A1R, EDA1R, EDA3, EDAR, HRM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10913 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UNE0 | |
| EDAR | |
| EDAR Fusion Protein Ag12601 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction