missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDA2R/TNFRSF27/XEDAR Rabbit anti-Human, Clone: 72, Novus Biologicals™
Rabbit Monoclonal Antibody
£286.00 - £429.00
Specifications
| Antigen | EDA2R/TNFRSF27/XEDAR |
|---|---|
| Clone | 72 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18256586
|
Novus Biologicals
NBP2-90000-50UL |
50 μg |
£286.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18286926
|
Novus Biologicals
NBP2-90000-100UL |
100 μg |
£429.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EDA2R/TNFRSF27/XEDAR Monoclonal specifically detects EDA2R/TNFRSF27/XEDAR in Human samples. It is validated for ELISA.Specifications
| EDA2R/TNFRSF27/XEDAR | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 60401 | |
| IgG | |
| Protein A purified |
| 72 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ectodysplasin A2 receptor, EDA-A2 receptor, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor | |
| Recombinant Human EDA2R/TNFRSF27/XEDAR protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title