missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDA2R/TNFRSF27/XEDAR Rabbit anti-Human, Clone: 72, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90000-50UL
This item is not returnable.
View return policy
Description
EDA2R/TNFRSF27/XEDAR Monoclonal specifically detects EDA2R/TNFRSF27/XEDAR in Human samples. It is validated for ELISA.
Specifications
| EDA2R/TNFRSF27/XEDAR | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 60401 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 72 | |
| ELISA 1:5000-1:10000 | |
| ectodysplasin A2 receptor, EDA-A2 receptor, EDA-A2R, EDAA2Rtumor necrosis factor receptor superfamily member XEDAR, EDAR2, XEDARTNFRSF27tumor necrosis factor receptor superfamily member 27, X-linked ectodysplasin-A2 receptor | |
| Recombinant Human EDA2R/TNFRSF27/XEDAR protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction