Learn More
Envair™ ecoChem Plus Recirculating Fume Cupboard
Provides fume containment and a safe working environment for the protection of laboratory personnel from chemicals vapors and aerosol.
Brand: Envair™ O00001870010
Code : NEW
Additional Details : Weight : 135.00000kg
Description
- Safety glass front screen – hinged, upward opening front window, designed for ergonomic instrument use in the work chamber, as well as easy access for cleaning and maintenance
- Built-in Safety – all electronic components, switches, lighting and motor fan have been installed in environments completely isolated from air contaminated with solvents
- Work surface supplied with acid resistant PVC specially designed to divert spilt liquids to the corners of the work surface
- Construction – cold rolled epoxy painted galvanized body for superior resistance to chemical solvents
- IP54 centrifugal motor fan, capable of maintaining a constant airflow by compensating for prefilter clogging, which occurs during normal operation
- Equipped with a microprocessor-based monitoring system, with LCD control panel showing the face speed and a digital system to select the most suitable velocity for each specific containment
- Charcoal filter is easily replaced from the front
- Large absorption capacity and filtration efficiency
- Applications include clinical diagnostic testing, biological and medical research, analytical chemistry, biotechnology, pharmaceutical industries and more
Specifications
Fume Cupboard | |
Microprocessor-based monitoring system | |
264 W |
>1000 lx | |
IP20 |